missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-trifluoromethoxy-DL-phenylalanine, 97%

Manufacturer:  Alfa Aesar™ H3403903

 View more versions of this product

Catalog No. AAH3403903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methoxy-3-trifluoromethoxy-DL-phenylalanine
C11H12F3NO4
4-methoxy-3-trifluoromethoxy-dl-phenylalanine, 2-amino-3-4-methoxy-3-trifluoromethoxy phenyl propanoic acid, 2-amino-3-4-methoxy-3-trifluoromethoxy phenyl propanoic acid, 4-methoxy-3-trifluoromethoxy-dl-phenylalanine
COC1=C(C=C(C=C1)CC(C(=O)O)N)OC(F)(F)F
279.215
279.21
97%
1259979-50-4
MFCD09915503
RAISOBRVUKCEEC-UHFFFAOYSA-N
2-amino-3-[4-methoxy-3-(trifluoromethoxy)phenyl]propanoic acid
66523527
Solid
1g

Chemical Identifiers

1259979-50-4
279.215
RAISOBRVUKCEEC-UHFFFAOYSA-N
66523527
COC1=C(C=C(C=C1)CC(C(=O)O)N)OC(F)(F)F
C11H12F3NO4
MFCD09915503
4-methoxy-3-trifluoromethoxy-dl-phenylalanine, 2-amino-3-4-methoxy-3-trifluoromethoxy phenyl propanoic acid, 2-amino-3-4-methoxy-3-trifluoromethoxy phenyl propanoic acid, 4-methoxy-3-trifluoromethoxy-dl-phenylalanine
2-amino-3-[4-methoxy-3-(trifluoromethoxy)phenyl]propanoic acid
SDS Documents