Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-trifluoromethoxy-DL-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-trifluoromethoxy-DL-phenylalanine
Formula Weight 279.21
MDL Number MFCD09915503
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3403903

  SDS
alfa aesar™
H3403903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH34039MD

  SDS
alfa aesar™
H34039MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 279.21
Chemical Name or Material 4-Methoxy-3-trifluoromethoxy-DL-phenylalanine
Physical Form Solid
Assay 97%
MDL Number MFCD09915503
Quantity 1g