Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-(trifluoromethoxy)benzaldehyde, 98%

$167.26 - $415.86

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-(trifluoromethoxy)benzaldehyde
CAS 853771-90-1
Molecular Formula C9H7F3O3
Formula Weight 220.15
MDL Number MFCD08062376
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2614003

  SDS
alfa aesar™
H2614003
1g Each for $415.86

AAH26140MD

  SDS
alfa aesar™
H26140MD
250mg Each for $167.26
Description & Specifications

Specifications

Formula Weight 220.15
Molecular Formula C9H7F3O3
Chemical Name or Material 4-Methoxy-3-(trifluoromethoxy)benzaldehyde
Assay 98%
MDL Number MFCD08062376
CAS 853771-90-1
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Methoxy-3-(trifluoromethoxy)benzaldehyde is used as intermediates in pharmaceutical industry.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.