Learn More
4-Methoxy-3-pyridineboronic acid hydrate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 378380010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Methoxy-3-pyridineboronic acid hydrate | |
Authentic | |
Glass bottle | |
MFCD01114676 | |
4-methoxy-3-pyridineboronic acid, 4-methoxypyridin-3-yl boronic acid, 4-methoxypyridine-3-boronic acid, 3-borono-4-methoxypyridine, 4-methoxy-3-pyridylboronic acid, boronic acid, 4-methoxy-3-pyridinyl, 4-methoxy-pyridine-3-boric acid, acmc-209iha, 4-methoxy-5-pyridineboronic acid, 4-methoxy-pyridin-3-boronic acid | |
COC1=C(C=NC=C1)B(O)O | |
152.94 | |
152.94 |
1266615-79-5,355004-67-0 | |
96% min. (HPLC) | |
C6H8BNO3 | |
1g | |
YUTPAZKVEOJQCY-UHFFFAOYSA-N | |
(4-methoxypyridin-3-yl)boronic acid | |
2762556 | |
97% |
Chemical Identifiers
1266615-79-5 | |
152.94 | |
YUTPAZKVEOJQCY-UHFFFAOYSA-N | |
2762556 | |
COC1=C(C=NC=C1)B(O)O |
C6H8BNO3 | |
MFCD01114676 | |
4-methoxy-3-pyridineboronic acid, 4-methoxypyridin-3-yl boronic acid, 4-methoxypyridine-3-boronic acid, 3-borono-4-methoxypyridine, 4-methoxy-3-pyridylboronic acid, boronic acid, 4-methoxy-3-pyridinyl, 4-methoxy-pyridine-3-boric acid, acmc-209iha, 4-methoxy-5-pyridineboronic acid, 4-methoxy-pyridin-3-boronic acid | |
(4-methoxypyridin-3-yl)boronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only