Learn More
Alfa Aesar™ 4-Methoxy-3-nitropyridine hydrochloride, 97%
Manufacturer: Alfa Aesar™ A1511014
Specifications
4-Methoxy-3-nitropyridine hydrochloride | |
25g | |
C6H7ClN2O3 | |
4-methoxy-3-nitropyridine hydrochloride, 3-nitro-4-methoxypyridine hydrochloride, 4-methoxy-3-nitropyridine hydrogen chloride, pubchem6689, acmc-1ae1i, 4methoxy-3-nitropyridine hydrochloride, 4-methoxy-3-nitro-pyridine hydrochloride, pyridine, 4-methoxy-3-nitro-, monohydrochloride | |
COC1=C(C=NC=C1)[N+](=O)[O-].Cl | |
190.583 | |
190.59 |
~270°C (decomposition) | |
31872-61-4 | |
MFCD00051995 | |
RRJVDMBZMAGZGE-UHFFFAOYSA-N | |
4-methoxy-3-nitropyridine;hydrochloride | |
355831 | |
97% |
Chemical Identifiers
31872-61-4 | |
190.583 | |
RRJVDMBZMAGZGE-UHFFFAOYSA-N | |
355831 | |
COC1=C(C=NC=C1)[N+](=O)[O-].Cl |
C6H7ClN2O3 | |
MFCD00051995 | |
4-methoxy-3-nitropyridine hydrochloride, 3-nitro-4-methoxypyridine hydrochloride, 4-methoxy-3-nitropyridine hydrogen chloride, pubchem6689, acmc-1ae1i, 4methoxy-3-nitropyridine hydrochloride, 4-methoxy-3-nitro-pyridine hydrochloride, pyridine, 4-methoxy-3-nitro-, monohydrochloride | |
4-methoxy-3-nitropyridine;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning