missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-nitropyridine hydrochloride, 97%

Manufacturer:  Alfa Aesar™ A1511003

 View more versions of this product

Catalog No. AAA1511003



Specifications

Specifications

4-Methoxy-3-nitropyridine hydrochloride
1g
C6H7ClN2O3
4-methoxy-3-nitropyridine hydrochloride, 3-nitro-4-methoxypyridine hydrochloride, 4-methoxy-3-nitropyridine hydrogen chloride, pubchem6689, acmc-1ae1i, 4methoxy-3-nitropyridine hydrochloride, 4-methoxy-3-nitro-pyridine hydrochloride, pyridine, 4-methoxy-3-nitro-, monohydrochloride
COC1=C(C=NC=C1)[N+](=O)[O-].Cl
190.583
190.59
~270°C (decomposition)
31872-61-4
MFCD00051995
RRJVDMBZMAGZGE-UHFFFAOYSA-N
4-methoxy-3-nitropyridine;hydrochloride
355831
97%

Chemical Identifiers

31872-61-4
190.583
RRJVDMBZMAGZGE-UHFFFAOYSA-N
355831
COC1=C(C=NC=C1)[N+](=O)[O-].Cl
C6H7ClN2O3
MFCD00051995
4-methoxy-3-nitropyridine hydrochloride, 3-nitro-4-methoxypyridine hydrochloride, 4-methoxy-3-nitropyridine hydrogen chloride, pubchem6689, acmc-1ae1i, 4methoxy-3-nitropyridine hydrochloride, 4-methoxy-3-nitro-pyridine hydrochloride, pyridine, 4-methoxy-3-nitro-, monohydrochloride
4-methoxy-3-nitropyridine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents