Please login to your online account to display your discounted pricing

4-Methoxy-3-nitrobenzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-nitrobenzoic acid
Name Note 98%
CAS 89-41-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7NO5
Linear Formula CH3OC6H3(NO2)CO2H
Formula Weight 197.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC187831000

  SDS
thermo fisher scientific
187831000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC187830250

  SDS
thermo fisher scientific
187830250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 197.15
Quantity 100g
Color Light yellow
Melting Point 189° to 194°C
EINECS Number 201-906-0
TSCA TSCA
Beilstein 10, 181
CAS Max % 100.0
Chemical Name or Material 4-Methoxy-3-nitrobenzoic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula CH3OC6H3(NO2)CO2H
CAS Min % 97.5
Name Note 98%
Molecular Formula C8H7NO5
MDL Number MFCD00007256
CAS 89-41-8