Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-nitrobenzoic acid, 98%

$43.55 - $43.55

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-nitrobenzoic acid
CAS 89-41-8
Molecular Formula C8H7NO5
Formula Weight 197.15
MDL Number MFCD00007256
Beilstein 2115073
Synonym 3-Nitro-p-anisic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1093014

  SDS
alfa aesar™
A1093014
25g Each for $43.55
Description & Specifications

Specifications

Formula Weight 197.15
Assay 98%
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Methoxy-3-nitrobenzoic acid
Melting Point 185° to 190°C
CAS 89-41-8
Beilstein 2115073
Molecular Formula C8H7NO5
MDL Number MFCD00007256
Health Hazard 1 Warning
TSCA TSCA
Synonym 3-Nitro-p-anisic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.