Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-nitrobenzamide, 97%

$84.82 - $1,223.49

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-nitrobenzamide
CAS 10397-58-7
Molecular Formula C8H8N2O3
Formula Weight 180.17
MDL Number MFCD00017136
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2490503

  SDS
alfa aesar™
B2490503
1g Each for $84.82

AAB2490506

  SDS
alfa aesar™
B2490506
5g Each for $298.34

AAB2490514

  SDS
alfa aesar™
B2490514
25g Each for $1,223.49
Description & Specifications

Specifications

Formula Weight 180.17
TSCA TSCA
Health Hazard 1 Warning
Chemical Name or Material 4-Methoxy-3-nitrobenzamide
Assay 97%
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H8N2O3
Melting Point 173° to 176°C
CAS 10397-58-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00017136
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.