Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Methoxy-3'-nitroacetophenone, 98+%

$97.43 - $357.11

Chemical Identifiers

Chemical Name or Material 4'-Methoxy-3'-nitroacetophenone
CAS 6277-38-9
Molecular Formula C9H9NO4
Formula Weight 195.18
MDL Number MFCD00017023
Beilstein 646069
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0414909

 
alfa aesar™
L0414909
10g Each for $97.43

AAL0414918

 
alfa aesar™
L0414918
50g Each for $357.11
Description & Specifications

Specifications

Formula Weight 195.18
Chemical Name or Material 4'-Methoxy-3'-nitroacetophenone
Assay ≥98%
Molecular Formula C9H9NO4
MDL Number MFCD00017023
CAS 6277-38-9
Beilstein 646069
Melting Point 97° to 99°C
Quantity 10g