Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Methoxy-3'-nitroacetophenone, 98+%


Quantity 50g

Manufacturer: alfa aesar™  L0414918

 View more versions of this product

Catalog No. AAL0414918

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 195.18
CAS 6277-38-9
MDL Number MFCD00017023
Assay ≥98%
Beilstein 646069
Quantity 50g
Melting Point 97° to 99°C
Molecular Formula C9H9NO4
Chemical Name or Material 4'-Methoxy-3'-nitroacetophenone

Specifications

Chemical Name or Material 4'-Methoxy-3'-nitroacetophenone
CAS 6277-38-9
Molecular Formula C9H9NO4
Formula Weight 195.18
MDL Number MFCD00017023
Beilstein 646069