missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-methylbenzyl alcohol

$121.18 - $421.85

Chemical Identifiers

CAS 114787-91-6
Molecular Formula C9H12O2
Formula Weight 152.19
MDL Number MFCD00060357
Chemical Name or Material 4-Methoxy-3-methylbenzyl alcohol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5034403 SDS Alfa Aesar™
H5034403
1g Each for $421.85
AAH50344MD SDS Alfa Aesar™
H50344MD
250mg Each for $121.18
Specifications Description & Specifications

Chemical Identifiers

114787-91-6
152.19
4-Methoxy-3-methylbenzyl alcohol
C9H12O2
MFCD00060357

Specifications

1g
C9H12O2
MFCD00060357
114787-91-6
152.19
4-Methoxy-3-methylbenzyl alcohol

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning