Please login to your online account to display your discounted pricing

4-Methoxy-3-methylbenzoic acid, +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-methylbenzoic acid
CAS 6880-04-2
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C9H10O3
Linear Formula CH3C6H3(OCH3)CO2H
Formula Weight 116.18
MDL Number MFCD00270114
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC394010010

  SDS
thermo fisher scientific
AC394010010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Powder
Formula Weight 116.18
Quantity 1g
Color Pale yellow or cream
Melting Point 194° to 198°C
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass bottle
Linear Formula CH3C6H3(OCH3)CO2H
Molecular Formula C9H10O3
MDL Number MFCD00270114
CAS 6880-04-2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Methoxy-3-methylbenzoic acid
Infrared Spectrum Authentic
Assay Percent Range 98+%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.