Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-methylbenzeneboronic acid, 98+%

$34.96 - $145.09

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-methylbenzeneboronic acid
CAS 175883-62-2
Molecular Formula C8H11BO3
Formula Weight 165.99
MDL Number MFCD02179464
Beilstein 7475212
Synonym 4-Methoxy-3-methylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1982103

  SDS
alfa aesar™
L1982103
1g Each for $34.96

AAL1982106

  SDS
alfa aesar™
L1982106
5g Each for $145.09
Description & Specifications

Specifications

Formula Weight 165.99
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
MDL Number MFCD02179464
Synonym 4-Methoxy-3-methylphenylboronic acid
Molecular Formula C8H11BO3
Chemical Name or Material 4-Methoxy-3-methylbenzeneboronic acid
CAS 175883-62-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Melting Point ≈215°C
Beilstein 7475212
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.