missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-(4-morpholinylsulfonyl)benzeneboronic acid, 95%

Manufacturer:  Alfa Aesar™ H5332903

 View more versions of this product

Catalog No. AAH5332903



Specifications

Specifications

4-Methoxy-3-(4-morpholinylsulfonyl)benzeneboronic acid
1g
C11H16BNO6S
4-methoxy-3-morpholinosulfonyl phenyl boronic acid, 4-methoxy-3-n-morpholinylsulfonyl phenylboronic acid, 4-methoxy-3-morpholin-4-ylsulphonyl benzeneboronic acid, 4-methoxy-3-morpholine-4-sulfonyl phenylboronic acid, boronic acid,b-3-methoxy-5-4-morpholinylsulfonyl phenyl, acmc-209qhv, 4-borono-2-morpholin-4-yl sulphonyl anisole, 4-5-borono-2-methoxyphenyl sulphonyl morpholine, 4-methoxy-3-morpholinosulfonyl phenylboronic acid, 4-methoxy-3-morpholine-4-sulfonyl phenyl boronic acid
B(C1=CC(=C(C=C1)OC)S(=O)(=O)N2CCOCC2)(O)O
301.12
301.12
270°C to 276°C
871333-02-7
MFCD08056366
MCYOYCLLZQYKDW-UHFFFAOYSA-N
(4-methoxy-3-morpholin-4-ylsulfonylphenyl)boronic acid
44119826
95%

Chemical Identifiers

871333-02-7
301.12
MCYOYCLLZQYKDW-UHFFFAOYSA-N
44119826
B(C1=CC(=C(C=C1)OC)S(=O)(=O)N2CCOCC2)(O)O
C11H16BNO6S
MFCD08056366
4-methoxy-3-morpholinosulfonyl phenyl boronic acid, 4-methoxy-3-n-morpholinylsulfonyl phenylboronic acid, 4-methoxy-3-morpholin-4-ylsulphonyl benzeneboronic acid, 4-methoxy-3-morpholine-4-sulfonyl phenylboronic acid, boronic acid,b-3-methoxy-5-4-morpholinylsulfonyl phenyl, acmc-209qhv, 4-borono-2-morpholin-4-yl sulphonyl anisole, 4-5-borono-2-methoxyphenyl sulphonyl morpholine, 4-methoxy-3-morpholinosulfonyl phenylboronic acid, 4-methoxy-3-morpholine-4-sulfonyl phenyl boronic acid
(4-methoxy-3-morpholin-4-ylsulfonylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents