Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-(trifluoromethyl)benzyl alcohol, 97%

$128.22 - $512.30

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzyl alcohol
Molecular Formula C9H9F3O2
Formula Weight 206.16
MDL Number MFCD06202983
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3165903

  SDS
alfa aesar™
H3165903
1g Each for $128.22

AAH3165906

  SDS
alfa aesar™
H3165906
5g Each for $512.30
Description & Specifications

Specifications

Formula Weight 206.16
Assay 97%
Molecular Formula C9H9F3O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzyl alcohol
Quantity 1g
Health Hazard 1 Warning
MDL Number MFCD06202983
Melting Point 42° to 44°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.