Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-(trifluoromethyl)benzaldehyde, 98%

$79.55 - $79.55

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzaldehyde
CAS 106312-36-1
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD01091011
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26123MD

  SDS
alfa aesar™
H26123MD
250mg Each for $79.55
Description & Specifications

Specifications

Formula Weight 204.15
Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzaldehyde
Assay 98%
Molecular Formula C9H7F3O2
MDL Number MFCD01091011
CAS 106312-36-1
Melting Point 39° to 41°C
Quantity 250mg
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.