Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-(trifluoromethoxy)benzoic acid, 97%

$104.42 - $311.91

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-(trifluoromethoxy)benzoic acid
Molecular Formula C9H7F3O4
Formula Weight 236.15
MDL Number MFCD06660285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2673703

  SDS
alfa aesar™
H2673703
1g Each for $104.42

AAH2673706

  SDS
alfa aesar™
H2673706
5g Each for $311.91
Description & Specifications

Specifications

Formula Weight 236.15
Health Hazard 1 Warning
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD06660285
Chemical Name or Material 4-Methoxy-2-(trifluoromethoxy)benzoic acid
Melting Point 99° to 101°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Molecular Formula C9H7F3O4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.