Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-(trifluoromethoxy)benzaldehyde, 97%

$146.77 - $447.51

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-(trifluoromethoxy)benzaldehyde
CAS 886503-52-2
Molecular Formula C9H7F3O3
Formula Weight 220.15
MDL Number MFCD06660317
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2636106

  SDS
alfa aesar™
H2636106
5g Each for $447.51

AAH2636103

  SDS
alfa aesar™
H2636103
1g Each for $146.77
Description & Specifications

Specifications

Formula Weight 220.15
Chemical Name or Material 4-Methoxy-2-(trifluoromethoxy)benzaldehyde
Assay 97%
Molecular Formula C9H7F3O3
MDL Number MFCD06660317
CAS 886503-52-2
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.