Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-nitrophenol, 98%

$55.55 - $166.24

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-nitrophenol
CAS 1568-70-3
Molecular Formula C7H7NO4
Formula Weight 169.14
MDL Number MFCD00024247
Synonym 4-Hydroxy-3-nitroanisole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1340909

 
alfa aesar™
A1340909
10g Each for $55.55

AAA1340918

 
alfa aesar™
A1340918
50g Each for $166.24
Description & Specifications

Specifications

Formula Weight 169.14
Chemical Name or Material 4-Methoxy-2-nitrophenol
Assay 98%
Molecular Formula C7H7NO4
MDL Number MFCD00024247
CAS 1568-70-3
Melting Point 77° to 80°C
Synonym 4-Hydroxy-3-nitroanisole
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol almost transparency.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.