missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxy-2-nitroaniline 98.0+%, TCI America™
Supplier: TCI America M0119100G
Specifications
4-Methoxy-2-nitroaniline | |
124°C | |
MFCD00007152 | |
2811 | |
QFMJFXFXQAFGBO-UHFFFAOYSA-N | |
4-methoxy-2-nitroaniline | |
66793 | |
168.15 | |
Crystalline Powder |
96-96-8 | |
C7H8N2O3 | |
100 g | |
2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt | |
COC1=CC(=C(C=C1)N)[N+](=O)[O-] | |
168.152 | |
CHEBI:48973 | |
≥98.0% (GC) |
Chemical Identifiers
96-96-8 | |
168.152 | |
QFMJFXFXQAFGBO-UHFFFAOYSA-N | |
66793 | |
4-methoxy-2-nitroaniline |
C7H8N2O3 | |
MFCD00007152 | |
2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt | |
CHEBI:48973 | |
COC1=CC(=C(C=C1)N)[N+](=O)[O-] |
Safety and Handling
EINECSNumber : (5)-2296
RTECSNumber : BY4415000
TSCA : Yes