missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-2-nitroaniline, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1167322

 View more versions of this product

Catalog No. AAA1167322



Description

Description

4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

Solubility
Slightly soluble in water. Soluble in hot methanol almost transparency.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications

Specifications

4-Methoxy-2-nitroaniline
126°C to 128°C
MFCD00007152
UN2811
2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt
QFMJFXFXQAFGBO-UHFFFAOYSA-N
4-methoxy-2-nitroaniline
66793
168.15
96-96-8
C7H8N2O3
100 g
880318
Slightly soluble in water. Soluble in hot methanol almost transparency.
COC1=CC(=C(C=C1)N)[N+](=O)[O-]
168.152
CHEBI:48973
99%

Chemical Identifiers

96-96-8
168.152
QFMJFXFXQAFGBO-UHFFFAOYSA-N
66793
4-methoxy-2-nitroaniline
C7H8N2O3
MFCD00007152
2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt
CHEBI:48973
COC1=CC(=C(C=C1)N)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 202-547-2

RTECSNumber : BY4415000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only