Learn More
4-Methoxy-2-nitroaniline, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 125961000
Specifications
4-Methoxy-2-nitroaniline | |
123.0°C to 126.0°C | |
98% | |
96.0 | |
96% min. (HPLC) | |
CH3OC6H3(NO2)NH2 | |
13, 521 | |
Solubility in water: slightly soluble | |
COC1=CC(=C(C=C1)N)[N+](=O)[O-] | |
168.15 | |
CHEBI:48973 | |
97% |
Glass bottle | |
100g | |
96-96-8 | |
100.0 | |
C7H8N2O3 | |
MFCD00007152 | |
2-nitro-4-methoxyaniline, 2-nitro-p-anisidine, 4-amino-3-nitroanisole, azobase nas, bordeaux gp base, bordeaux gp salt, devol bordeaux b, fast bordeaux gp, fast bordeaux gp base, fast bordeaux gp salt | |
QFMJFXFXQAFGBO-UHFFFAOYSA-N | |
4-methoxy-2-nitroaniline | |
66793 | |
168.15 |
Chemical Identifiers
96-96-8 | |
168.15 | |
QFMJFXFXQAFGBO-UHFFFAOYSA-N | |
66793 | |
4-methoxy-2-nitroaniline |
C7H8N2O3 | |
MFCD00007152 | |
2-nitro-4-methoxyaniline, 2-nitro-p-anisidine, 4-amino-3-nitroanisole, azobase nas, bordeaux gp base, bordeaux gp salt, devol bordeaux b, fast bordeaux gp, fast bordeaux gp base, fast bordeaux gp salt | |
CHEBI:48973 | |
COC1=CC(=C(C=C1)N)[N+](=O)[O-] |
Safety and Handling
GHS H Statement:
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-547-2
RTECSNumber : BY4415000
TSCA : TSCA