Learn More
4-Methoxy-2-methylphenylhydrazine hydrochloride, 96%, Thermo Scientific Chemicals
$72.55 - $211.44
Chemical Identifiers
CAS | 93048-16-9 |
---|---|
Molecular Formula | C8H13ClN2O |
Molecular Weight (g/mol) | 188.655 |
MDL Number | MFCD03426021 |
InChI Key | OIFKWIPSJGNUCC-UHFFFAOYSA-N |
Synonym | 4-methoxy-2-methylphenylhydrazine hydrochloride, 4-methoxy-2-methylphenyl hydrazine hydrochloride, 1-4-methoxy-2-methylphenyl hydrazine hydrochloride, 4-hydrazino-3-methylanisole hydrochloride, 4-methoxy-2-methylphenylhydrazinehydrochloride, 4-methoxy-2-methylphenyl-hydrazine hydrochloride, hydrazine, 4-methoxy-2-methylphenyl-, monohydrochloride |
PubChem CID | 21997741 |
IUPAC Name | (4-methoxy-2-methylphenyl)hydrazine;hydrochloride |
SMILES | CC1=C(C=CC(=C1)OC)NN.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1481003
|
Thermo Scientific Chemicals
L1481003 |
1 g |
Each for $72.55
|
|
|||||
AAL1481006
|
Thermo Scientific Chemicals
L1481006 |
5 g |
Each for $211.44
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93048-16-9 | |
188.655 | |
OIFKWIPSJGNUCC-UHFFFAOYSA-N | |
21997741 | |
CC1=C(C=CC(=C1)OC)NN.Cl |
C8H13ClN2O | |
MFCD03426021 | |
4-methoxy-2-methylphenylhydrazine hydrochloride, 4-methoxy-2-methylphenyl hydrazine hydrochloride, 1-4-methoxy-2-methylphenyl hydrazine hydrochloride, 4-hydrazino-3-methylanisole hydrochloride, 4-methoxy-2-methylphenylhydrazinehydrochloride, 4-methoxy-2-methylphenyl-hydrazine hydrochloride, hydrazine, 4-methoxy-2-methylphenyl-, monohydrochloride | |
(4-methoxy-2-methylphenyl)hydrazine;hydrochloride |
Specifications
93048-16-9 | |
C8H13ClN2O | |
1 g | |
7227704 | |
OIFKWIPSJGNUCC-UHFFFAOYSA-N | |
(4-methoxy-2-methylphenyl)hydrazine;hydrochloride | |
21997741 | |
96% |
∽110°C (decomposition) | |
MFCD03426021 | |
UN2811 | |
4-methoxy-2-methylphenylhydrazine hydrochloride, 4-methoxy-2-methylphenyl hydrazine hydrochloride, 1-4-methoxy-2-methylphenyl hydrazine hydrochloride, 4-hydrazino-3-methylanisole hydrochloride, 4-methoxy-2-methylphenylhydrazinehydrochloride, 4-methoxy-2-methylphenyl-hydrazine hydrochloride, hydrazine, 4-methoxy-2-methylphenyl-, monohydrochloride | |
CC1=C(C=CC(=C1)OC)NN.Cl | |
188.655 | |
188.66 | |
4-Methoxy-2-methylphenylhydrazine hydrochloride |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only