Learn More
4-Methoxy-2-methylphenylboronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 393850050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Methoxy-2-methylphenylboronic acid | |
96.0 | |
Authentic | |
Glass Bottle | |
MFCD02684315 | |
4-methoxy-2-methylphenyl boronic acid, 2-methyl-4-methoxyphenylboronic acid, 4-methoxy-2-methylbenzeneboronic acid, 4-methoxy-2-methylphenyl boranediol, 4-methoxy-2-methyl-phenyl boronic acid, 4-methoxy-2-methylphenbylboronic acid, 4-methoxy-2-methylphenylboronicacid, boronic acid, 4-methoxy-2-methylphenyl, 2-methyl-4-methoxybenzeneboronic acid, acmc-1cge6 | |
B(C1=C(C=C(C=C1)OC)C)(O)O | |
165.98 | |
165.98 |
208399-66-0 | |
100.0 | |
96% min. (HPLC) | |
C8H11BO3 | |
5g | |
AMSQNQJCBXQYEX-UHFFFAOYSA-N | |
(4-methoxy-2-methylphenyl)boronic acid | |
2773486 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
RUO â Research Use Only