Please login to your online account to display your discounted pricing

4-Methoxy-2-methylphenyl isocyanate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC377730050

 View more versions of this product

Catalog No. AC377730050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 163.17
Quantity 5g
Color Clear colorless to light yellow
Density 1.2100g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS 60385-06-0
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Refractive Index 1.5435 to 1.5465 (20°C, 589nm)
Chemical Name or Material 4-Methoxy-2-methylphenyl isocyanate
Packaging Glass bottle
Molecular Formula C9 H9 N O2
Linear Formula CH3C6H3(OCH3)NCO
MDL Number MFCD00013852
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Methoxy-2-methylphenyl isocyanate
CAS 60385-06-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H9 N O2
Linear Formula CH3C6H3(OCH3)NCO
Formula Weight 163.17
MDL Number MFCD00013852
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

SDS & Safety Documents