Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-methylphenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-methylphenyl isocyanate
CAS 60385-06-0
Molecular Formula C9H9NO2
Formula Weight 163.18
MDL Number MFCD00013852
UN Number UN2206
Beilstein 7115803
Synonym 4-Methoxy-o-tolyl isocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1208603

  SDS
alfa aesar™
L1208603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1208606

  SDS
alfa aesar™
L1208606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 163.18
Chemical Name or Material 4-Methoxy-2-methylphenyl isocyanate
CAS 60385-06-0
Health Hazard 1 Danger
Synonym 4-Methoxy-o-tolyl isocyanate
Molecular Formula C9H9NO2
Beilstein 7115803
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00013852
UN Number UN2206
Quantity 1g
Recommended Storage 2° to 8°C
Assay 97%
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Flash Point >110°C (230°F)
Boiling Point 74° to 76°C (1mmHg)
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C