missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-methylphenyl isocyanate, 97%

Manufacturer:  Alfa Aesar™ L1208606

 View more versions of this product

Catalog No. AAL1208606



Specifications

Specifications

4-Methoxy-2-methylphenyl isocyanate
74°C to 76°C (1mmHg)
60385-06-0
MFCD00013852
7115803
XGRQPTXFVDRJMY-UHFFFAOYSA-N
1-isocyanato-4-methoxy-2-methylbenzene
2734905
97%
>110°C (230°F)
5g
C9H9NO2
UN2206
4-methoxy-2-methylphenyl isocyanate, 4-methoxy-2-methylphenylisocyanate, benzene, 1-isocyanato-4-methoxy-2-methyl, acmc-1bfcu, 4-methoxy-o-tolyl isocyanate, 4-methoxy-2-methylbenzenisocyanate, 1-isocyanato-4-methoxy-2-methyl benzene, 1-isocyanato-4-methoxy-2-methyl-benzene, 4-methoxy-2-methylphenyl isocyanate, benzene,1-isocyanato-4-methoxy-2-methyl
CC1=C(C=CC(=C1)OC)N=C=O
163.176
163.18

Chemical Identifiers

60385-06-0
163.176
XGRQPTXFVDRJMY-UHFFFAOYSA-N
2734905
CC1=C(C=CC(=C1)OC)N=C=O
C9H9NO2
MFCD00013852
4-methoxy-2-methylphenyl isocyanate, 4-methoxy-2-methylphenylisocyanate, benzene, 1-isocyanato-4-methoxy-2-methyl, acmc-1bfcu, 4-methoxy-o-tolyl isocyanate, 4-methoxy-2-methylbenzenisocyanate, 1-isocyanato-4-methoxy-2-methyl benzene, 1-isocyanato-4-methoxy-2-methyl-benzene, 4-methoxy-2-methylphenyl isocyanate, benzene,1-isocyanato-4-methoxy-2-methyl
1-isocyanato-4-methoxy-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents