missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-2-methyldiphenylamine, TCI America™

$59.58 - $421.67

Chemical Identifiers

CAS 41317-15-1
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00272619
InChI Key CYMPUOGZUXAIMY-UHFFFAOYSA-N
PubChem CID 162461
IUPAC Name 4-methoxy-2-methyl-N-phenylaniline
SMILES CC1=C(C=CC(=C1)OC)NC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M128625G View Documents TCI America
M128625G
25g Each for $59.58
 
M1286500G View Documents TCI America
M1286500G
500g Each for $421.67
 
Specifications

Chemical Identifiers

41317-15-1
213.28
CYMPUOGZUXAIMY-UHFFFAOYSA-N
4-methoxy-2-methyl-N-phenylaniline
C14H15NO
MFCD00272619
162461
CC1=C(C=CC(=C1)OC)NC2=CC=CC=C2

Specifications

171°C
81°C
41317-15-1
MFCD00272619
CC1=C(C=CC(=C1)OC)NC2=CC=CC=C2
213.28
213.28
≥97.0% (GC)
Undesignated
25g
C14H15NO
CYMPUOGZUXAIMY-UHFFFAOYSA-N
4-methoxy-2-methyl-N-phenylaniline
162461
Crystal-Powder at 20°C
4-Methoxy-2-methyldiphenylamine
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3869

TSCA : Yes