missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-2-methylbenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393980010

 View more versions of this product

Catalog No. AC393980010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

166.18
96.0
6245-57-4
MFCD00020291
97%
CH3C6H3(OCH3)CO2H
1g
100.0
4-Methoxy-2-methylbenzoic acid
C9H10O3
Glass bottle

Chemical Identifiers

6245-57-4
100.0
C9H10O3
166.18
4-Methoxy-2-methylbenzoic acid
96.0
97%
CH3C6H3(OCH3)CO2H
MFCD00020291

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.