Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-methylbenzeneboronic acid, 98%

$94.47 - $322.45

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-methylbenzeneboronic acid
CAS 208399-66-0
Molecular Formula C8H11BO3
Formula Weight 165.99
MDL Number MFCD02684315
Beilstein 8111222
Synonym 4-Methoxy-2-methylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2011203

  SDS
alfa aesar™
L2011203
1g Each for $94.47

AAL2011206

  SDS
alfa aesar™
L2011206
5g Each for $322.45
Description & Specifications

Specifications

Formula Weight 165.99
Chemical Name or Material 4-Methoxy-2-methylbenzeneboronic acid
Assay 98%
MDL Number MFCD02684315
CAS 208399-66-0
Beilstein 8111222
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H11BO3
Synonym 4-Methoxy-2-methylphenylboronic acid
Melting Point 191°C
Quantity 1g

Applications
Reactant for preparation of biologically active molecule preparation of hydroxyphenylnaphthols as 17ß-hydroxysteroid dehydrogenase Type 2 inhibitors

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.