Please login to your online account to display your discounted pricing

4-Methoxy-2-methylbenzaldehyde, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC427020050

 View more versions of this product

Catalog No. AC427020050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 150.18
Quantity 5g
Color Clear colorless to yellow
Density 1.1070g/mL
CAS Min % 96.0
CAS Max % 100.0
Linear Formula CH30C6H3(CH3)CHO
Chemical Name or Material 4-Methoxy-2-methylbenzaldehyde
Assay Percent Range 97%
Molecular Formula C9H10O2
MDL Number MFCD02261771
CAS 52289-54-0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
NMR Consistent with structure

Specifications

Chemical Name or Material 4-Methoxy-2-methylbenzaldehyde
CAS 52289-54-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10O2
Linear Formula CH30C6H3(CH3)CHO
Formula Weight 150.18
MDL Number MFCD02261771
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.