Please login to your online account to display your discounted pricing

4-Methoxy-2-methylaniline 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-methylaniline
Name Note 98%
CAS 102-50-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H11NO
Linear Formula CH3OC6H3(CH3)NH2
Formula Weight 137.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC125910250

  SDS
thermo fisher scientific
125910250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC125910050

  SDS
thermo fisher scientific
125910050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 137.18
Quantity 25g
Color Brown to black
Beilstein 13, II, 330
EINECS Number 203-036-7
TSCA TSCA
RTECS Number BZ6730000
Density 1.0600g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
CAS 102-50-1
Synonym 2-Methyl-p-anisidine
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5670 to 1.5700 (20°C, 589nm)
Chemical Name or Material 4-Methoxy-2-methylaniline
Molecular Formula C8H11NO
Linear Formula CH3OC6H3(CH3)NH2
MDL Number MFCD00007735
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.