Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2-methylaniline, 98%

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2-methylaniline
CAS 102-50-1
Molecular Formula C8H11NO
Formula Weight 137.18
MDL Number MFCD00007735
UN Number UN2810
Beilstein 774727
Synonym m-Cresidine; 4-Methoxy-o-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1491006

  SDS
alfa aesar™
L1491006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1491014

  SDS
alfa aesar™
L1491014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1491022

  SDS
alfa aesar™
L1491022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 137.18
Molecular Formula C8H11NO
Health Hazard 3 GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2810
MDL Number MFCD00007735
TSCA TSCA
Beilstein 774727
Chemical Name or Material 4-Methoxy-2-methylaniline
Health Hazard 2 GHS H Statement
H331-H351-H302-H312
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
CAS 102-50-1
Melting Point 13° to 14°C
Flash Point 124°C (255°F)
Quantity 5g
Synonym m-Cresidine; 4-Methoxy-o-toluidine
Boiling Point 248° to 249°C
Health Hazard 1 Danger
RTECS Number BZ6730000
Assay 98%
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H351-H302-H312
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.