missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2,6-dimethylbenzeneboronic acid, 95%

Manufacturer:  Alfa Aesar™ H53355MD

 View more versions of this product

Catalog No. AAH53355MD



Description

Description

Applications
4-Methoxy-2,6-dimethylbenzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-Methoxy-2,6-dimethylbenzeneboronic acid
250mg
C9H13BO3
2,6-dimethyl-4-methoxyphenyl boronic acid, 2,6-dimethyl-4-methoxyphenylboronic acid, 4-methoxy-2,6-dimethylphenyl boronic acid, 2,6-dimethyl-4-methoxybenzeneboronic acid, 2-borono-5-methoxy-m-xylene, 4-borono-3,5-dimethylanisole, 4-methoxy-2,6-dimethylbenzeneboronic acid, 4-methoxy-2,6-dimethylphenylboronicacid, boronic acid, 4-methoxy-2,6-dimethylphenyl, acmc-209il9
B(C1=C(C=C(C=C1C)OC)C)(O)O
180.01
180.01
168°C to 172°C
361543-99-9
MFCD03452755
UOSSBXMFWPYHEF-UHFFFAOYSA-N
(4-methoxy-2,6-dimethylphenyl)boronic acid
3716303
95%

Chemical Identifiers

361543-99-9
180.01
UOSSBXMFWPYHEF-UHFFFAOYSA-N
3716303
B(C1=C(C=C(C=C1C)OC)C)(O)O
C9H13BO3
MFCD03452755
2,6-dimethyl-4-methoxyphenyl boronic acid, 2,6-dimethyl-4-methoxyphenylboronic acid, 4-methoxy-2,6-dimethylphenyl boronic acid, 2,6-dimethyl-4-methoxybenzeneboronic acid, 2-borono-5-methoxy-m-xylene, 4-borono-3,5-dimethylanisole, 4-methoxy-2,6-dimethylbenzeneboronic acid, 4-methoxy-2,6-dimethylphenylboronicacid, boronic acid, 4-methoxy-2,6-dimethylphenyl, acmc-209il9
(4-methoxy-2,6-dimethylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents