Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride, tech. 90%

Chemical Identifiers

Chemical Name or Material 4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride
CAS 80745-07-9
Purity Grade Technical Grade
Molecular Formula C10H13ClO3S
Formula Weight 248.73
MDL Number MFCD00038846
UN Number UN3261
Beilstein 4867665
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1182906

 
alfa aesar™
L1182906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1182903

 
alfa aesar™
L1182903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.73
Chemical Name or Material 4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride
Purity Grade Technical Grade
Molecular Formula C10H13ClO3S
MDL Number MFCD00038846
CAS 80745-07-9
UN Number UN3261
Melting Point 56° to 59°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 90%
Beilstein 4867665
Quantity 5g
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

−30° to −10°C