missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride, tech. 90%

Manufacturer:  Alfa Aesar™ L1182903

 View more versions of this product

Catalog No. AAL1182903



Specifications

Specifications

4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride
1g
C10H13ClO3S
UN3261
4-methoxy-2,3,6-trimethylbenzene-1-sulfonyl chloride, 4-methoxy-2,3,6-trimethyl-benzenesulfonyl chloride, mtr chloride, 4-methoxy-2,3,6-trimethylbenzenesulfonylchloride, 4-methoxy-2,3,6-trimethylphenylsulfonylchloride, chloro 4-methoxy-2,3,6-trimethylphenyl sulfone, 4-methoxy-2,3,6-trimethylphenyl sulfonylchloride, 4-methoxy-2,3,6-trimethylbenzensulfonyl chloride, 4-methoxy-2,3,6-trimethylphenylaulfomyl chloride, 4-methoxy-2,3,6-trimethylphenylsulfonyl chloride
CC1=CC(=C(C(=C1S(=O)(=O)Cl)C)C)OC
248.721
248.73
Technical
56°C to 59°C
80745-07-9
MFCD00038846
4867665
DWLYVEYCCPEHLX-UHFFFAOYSA-N
4-methoxy-2,3,6-trimethylbenzenesulfonyl chloride
562787
90%

Chemical Identifiers

80745-07-9
248.721
DWLYVEYCCPEHLX-UHFFFAOYSA-N
562787
CC1=CC(=C(C(=C1S(=O)(=O)Cl)C)C)OC
C10H13ClO3S
MFCD00038846
4-methoxy-2,3,6-trimethylbenzene-1-sulfonyl chloride, 4-methoxy-2,3,6-trimethyl-benzenesulfonyl chloride, mtr chloride, 4-methoxy-2,3,6-trimethylbenzenesulfonylchloride, 4-methoxy-2,3,6-trimethylphenylsulfonylchloride, chloro 4-methoxy-2,3,6-trimethylphenyl sulfone, 4-methoxy-2,3,6-trimethylphenyl sulfonylchloride, 4-methoxy-2,3,6-trimethylbenzensulfonyl chloride, 4-methoxy-2,3,6-trimethylphenylaulfomyl chloride, 4-methoxy-2,3,6-trimethylphenylsulfonyl chloride
4-methoxy-2,3,6-trimethylbenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : −30° to −10°C

Documents