Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-1-naphthonitrile, 98%

Chemical Identifiers

Chemical Name or Material 4-Methoxy-1-naphthonitrile
CAS 5961-55-7
Molecular Formula C12H9NO
Formula Weight 183.21
MDL Number MFCD00003925
UN Number UN3439
Beilstein 2096262
Synonym 1-Cyano-4-methoxynaphthalene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0535414

  SDS
alfa aesar™
L0535414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0535406

  SDS
alfa aesar™
L0535406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.21
Molecular Formula C12H9NO
CAS 5961-55-7
Chemical Name or Material 4-Methoxy-1-naphthonitrile
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-Cyano-4-methoxynaphthalene
Assay 98%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Beilstein 2096262
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 100° to 103°C
Quantity 25g
MDL Number MFCD00003925
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.