missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-1-naphthalenemethanol, TCI America™

$90.14 - $265.84

Chemical Identifiers

CAS 16820-54-5
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD04089264
InChI Key VBXSDYNXURBEQH-UHFFFAOYSA-N
Synonym 1-(Hydroxymethyl)-4-methoxynaphthalene
PubChem CID 900419
IUPAC Name (4-methoxynaphthalen-1-yl)methanol
SMILES COC1=CC=C(C2=CC=CC=C21)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20861G View Documents TCI America
M20861G
1g Each for $90.14
 
M20865G View Documents TCI America
M20865G
5g Each for $265.84
 
Specifications

Chemical Identifiers

16820-54-5
188.226
VBXSDYNXURBEQH-UHFFFAOYSA-N
900419
COC1=CC=C(C2=CC=CC=C21)CO
C12H12O2
MFCD04089264
1-(Hydroxymethyl)-4-methoxynaphthalene
(4-methoxynaphthalen-1-yl)methanol

Specifications

Undesignated
1g
C12H12O2
1-(Hydroxymethyl)-4-methoxynaphthalene
COC1=CC=C(C2=CC=CC=C21)CO
188.226
188.23
≥96.0% (GC)
76°C
16820-54-5
MFCD04089264
VBXSDYNXURBEQH-UHFFFAOYSA-N
(4-methoxynaphthalen-1-yl)methanol
900419
Crystal-Powder at 20°C
4-Methoxy-1-naphthalenemethanol
Safety and Handling

Safety and Handling

TSCA : No