Please login to your online account to display your discounted pricing

4-Methoxy-1-naphthaldehyde 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  146940100

 View more versions of this product

Catalog No. AC146940100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Low Melting Solid
Formula Weight 186.21
Quantity 10g
Color Slightly beige to pink-brown
Beilstein 08, 147
EINECS Number 240-109-2
Melting Point 32° to 35°C
CAS Max % 100.0
Chemical Name or Material 4-Methoxy-1-naphthaldehyde
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Linear Formula CH3OC10H6CHO
CAS 15971-29-6
CAS Min % 98.5
Molecular Formula C12H10O2
MDL Number MFCD00004006

Specifications

Chemical Name or Material 4-Methoxy-1-naphthaldehyde
Name Note 99%
CAS 15971-29-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H10O2
Linear Formula CH3OC10H6CHO
Formula Weight 186.21
MDL Number MFCD00004006