Please login to your online account to display your discounted pricing

4-Methoxy-1-naphthaldehyde 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methoxy-1-naphthaldehyde
Name Note 99%
CAS 15971-29-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H10O2
Linear Formula CH3OC10H6CHO
Formula Weight 186.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146940100

  SDS
fisher scientific acros
146940100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Low Melting Solid
Formula Weight 186.21
Quantity 10g
Color Slightly beige to pink-brown
Beilstein 08, 147
EINECS Number 240-109-2
Melting Point 32° to 35°C
CAS Max % 100.0
Chemical Name or Material 4-Methoxy-1-naphthaldehyde
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Linear Formula CH3OC10H6CHO
CAS 15971-29-6
CAS Min % 98.5
Molecular Formula C12H10O2
MDL Number MFCD00004006