Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-1-naphthaldehyde, 99%

$85.97 - $770.95

Chemical Identifiers

Chemical Name or Material 4-Methoxy-1-naphthaldehyde
CAS 15971-29-6
Molecular Formula C12H10O2
Formula Weight 186.21g/mol
MDL Number MFCD00004006
Beilstein 2046436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1109406

 
alfa aesar™
A11094-06
5g Each for $85.97

AAA1109414

 
alfa aesar™
A11094-14
25g Each for $255.41

AAA1109422

 
alfa aesar™
A11094-22
100g Each for $770.95
Description & Specifications

Specifications

Quantity 5g
Melting Point 32° to 36°C
Boiling Point 154° to 156°C (1mmHg)
Quantity 5g
Flash Point >110°C (230°F)
Chemical Name or Material 4-Methoxy-1-naphthaldehyde
CAS 15971-29-6
Assay 99%
Molecular Formula C12H10O2
Formula Weight 186.21g/mol
MDL Number MFCD00004006
Beilstein 2046436