Please login to your online account to display your discounted pricing

4-Methoxy-1,2-benzisoxazol-3-amine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Methoxy-1, 2-benzisoxazol-3-amine
CAS 177995-40-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8N2O2
Formula Weight 164.16
MDL Number MFCD03407354
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00345CB

 
fisher scientific company
MO00345CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.16
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-Methoxy-1, 2-benzisoxazol-3-amine
Assay Percent Range 97%
Molecular Formula C8H8N2O2
MDL Number MFCD03407354
Packaging Amber Glass Bottle
CAS 177995-40-3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.