Learn More
4-(Methanesulfonyl)phenylboronic acid, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC359510050
Specifications
4-(Methanesulfonyl)phenylboronic acid | |
Glass bottle | |
275°C to 277°C | |
149104-88-1 | |
100.0 | |
C7H9BO4S | |
VDUKDQTYMWUSAC-UHFFFAOYSA-N | |
(4-methylsulfonylphenyl)boronic acid | |
2734364 | |
Powder |
Authentic | |
White | |
5g | |
98.0 | |
98% min. (HPLC) | |
4-boronophenyl methyl sulphone, 4-methanesulfonyl benzeneboronic acid, 4-methanesulfonyl phenylboronic acid, 4-methanesulfonylphenyl boranediol, 4-methanesulfonylphenylboronic acid, 4-methylsulfonyl phenyl boronic acid, 4-methylsulfonyl phenylboronic acid, 4-methylsulfonylphenyl boronic acid, 4-methylsulphonyl benzeneboronic acid, 4-methylsulphonyl phenylboronic acid | |
B(C1=CC=C(C=C1)S(=O)(=O)C)(O)O | |
200.02 | |
200.02 | |
98+% |
Chemical Identifiers
149104-88-1 | |
200.02 | |
4-boronophenyl methyl sulphone, 4-methanesulfonyl benzeneboronic acid, 4-methanesulfonyl phenylboronic acid, 4-methanesulfonylphenyl boranediol, 4-methanesulfonylphenylboronic acid, 4-methylsulfonyl phenyl boronic acid, 4-methylsulfonyl phenylboronic acid, 4-methylsulfonylphenyl boronic acid, 4-methylsulphonyl benzeneboronic acid, 4-methylsulphonyl phenylboronic acid | |
(4-methylsulfonylphenyl)boronic acid |
C7H9BO4S | |
VDUKDQTYMWUSAC-UHFFFAOYSA-N | |
2734364 | |
B(C1=CC=C(C=C1)S(=O)(=O)C)(O)O |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning