Please login to your online account to display your discounted pricing

4-(Methanesulfonamido)phenylboronic acid pinacol ester, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-(Methanesulfonamido)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C13H20BNO4S
Formula Weight 297.18
Synonym N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-phenyl]methanesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC445100050

  SDS
thermo fisher scientific
AC445100050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC445100010

  SDS
thermo fisher scientific
AC445100010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-(Methanesulfonamido)phenylboronic acid pinacol ester
CAS 616880-14-9
Assay Percent Range 97%
Molecular Formula C13H20BNO4S
Formula Weight 297.18
Synonym N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-phenyl]methanesulfonamide
Quantity 5g