missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methanesulfonamido)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445100010

 View more versions of this product

Catalog No. AC445100010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-(Methanesulfonamido)phenylboronic acid pinacol ester
196°C to 197°C
616880-14-9
C13H20BNO4S
NPSPNWPZDXQYAC-UHFFFAOYSA-N
N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulfonamide
16217653
97%
Glass Bottle
1g
96% min. (GC)
4-methanesulfonamido phenylboronic acid pinacol ester, 4-methanesulfonylamino phenyl boronic acid pinacol ester, 4-methanesulfonylamino phenylboronic acid pinacol ester, 4-methanesulfonylaminophenylboronic acid pinacol ester, 4-methanesulfonylaminophenylboronic acid, pinacol ester, amtb416, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanesulfonamide, n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanesulfonamide
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NS(=O)(=O)C
297.18
297.18

Chemical Identifiers

616880-14-9
297.18
4-methanesulfonamido phenylboronic acid pinacol ester, 4-methanesulfonylamino phenyl boronic acid pinacol ester, 4-methanesulfonylamino phenylboronic acid pinacol ester, 4-methanesulfonylaminophenylboronic acid pinacol ester, 4-methanesulfonylaminophenylboronic acid, pinacol ester, amtb416, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanesulfonamide, n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanesulfonamide
N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulfonamide
C13H20BNO4S
NPSPNWPZDXQYAC-UHFFFAOYSA-N
16217653
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NS(=O)(=O)C
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents