missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Mercaptopyridine, 96%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC164710500

 View more versions of this product

Catalog No. AC164710500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Mercaptopyridine
Glass bottle
182°C to 189°C
4556-23-4
100.0
C5H5NS
MFCD00006422
4 1h-pyridinethione
FHTDDANQIMVWKZ-UHFFFAOYSA-N
1H-pyridine-4-thione
2723889
Crystalline Powder
Authentic
Brown or Green-Yellow to Yellow
50g
95.0
96%
(C5H4N)SH
21, II, 35
Solubility in water: slightly soluble. Other solubilities: soluble in acetone,toluene,methanol and ethanol
C1=CNC=CC1=S
111.16
111.16
96%

Chemical Identifiers

4556-23-4
111.16
FHTDDANQIMVWKZ-UHFFFAOYSA-N
2723889
C1=CNC=CC1=S
C5H5NS
MFCD00006422
4 1h-pyridinethione
1H-pyridine-4-thione
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

EINECSNumber : 224-926-1

SDS Documents