Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Mercaptophenylacetic acid, 97%

$107.43 - $351.08

Chemical Identifiers

Chemical Name or Material 4-Mercaptophenylacetic acid
CAS 39161-84-7
Molecular Formula C8H8O2S
Formula Weight 168.21
MDL Number MFCD00797617
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2765803

  SDS
alfa aesar™
H2765803
1g Each for $107.43

AAH2765806

  SDS
alfa aesar™
H2765806
5g Each for $351.08
Description & Specifications

Specifications

Formula Weight 168.21
Chemical Name or Material 4-Mercaptophenylacetic acid
Assay 97%
Molecular Formula C8H8O2S
MDL Number MFCD00797617
CAS 39161-84-7
Health Hazard 1 Warning
Melting Point 105° to 110°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Recommended Storage 2° to 8°C

Applications
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.

Solubility
Slightly soluble in water. Soluble in Dichloromethane, Ethanol and Methanol

Notes
Air sensitive. Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C