missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Maleimidobutyric Acid, TCI America™

$93.20 - $323.89

Chemical Identifiers

CAS 57078-98-5
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.163
MDL Number MFCD00043139
InChI Key NCPQROHLJFARLL-UHFFFAOYSA-N
PubChem CID 3404904
IUPAC Name 4-(2,5-dioxopyrrol-1-yl)butanoic acid
SMILES C1=CC(=O)N(C1=O)CCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23371G View Documents TCI America
M23371G
1g Each for $93.20
 
M23375G View Documents TCI America
M23375G
5g Each for $323.89
 
Specifications

Chemical Identifiers

57078-98-5
183.163
NCPQROHLJFARLL-UHFFFAOYSA-N
4-(2,5-dioxopyrrol-1-yl)butanoic acid
C8H9NO4
MFCD00043139
3404904
C1=CC(=O)N(C1=O)CCCC(=O)O

Specifications

White-Yellow
1g
C8H9NO4
NCPQROHLJFARLL-UHFFFAOYSA-N
4-(2,5-dioxopyrrol-1-yl)butanoic acid
3404904
Crystal-Powder at 20°C
4-Maleimidobutyric Acid
96°C
57078-98-5
MFCD00043139
C1=CC(=O)N(C1=O)CCCC(=O)O
183.163
183.16
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator