missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(m-Tolylamino)pyridine-3-sulfonamide, TCI America™

Manufacturer:  TCI America T300725G

 View more versions of this product

Catalog No. T300725G



Specifications

Specifications

4-(m-Tolylamino)pyridine-3-sulfonamide
170°C
72811-73-5
MFCD00661332
ZXPCUGWAKUIOOF-UHFFFAOYSA-N
4-(3-methylanilino)pyridine-3-sulfonamide
819349
Crystal-Powder at 20°C
White
25g
C12H13N3O2S
4-(3-Methylphenyl)aminopyridine-3-sulfonamide, 4-(m-Tolylamino)-3-sulfamoylpyridine, 4-(3-Methylphenyl)amino-3-sulfamoylpyridine
CC1=CC(=CC=C1)NC2=C(C=NC=C2)S(=O)(=O)N
263.315
263.32
≥98.0% (HPLC,T)

Chemical Identifiers

72811-73-5
263.315
ZXPCUGWAKUIOOF-UHFFFAOYSA-N
819349
CC1=CC(=CC=C1)NC2=C(C=NC=C2)S(=O)(=O)N
C12H13N3O2S
MFCD00661332
4-(3-Methylphenyl)aminopyridine-3-sulfonamide, 4-(m-Tolylamino)-3-sulfamoylpyridine, 4-(3-Methylphenyl)amino-3-sulfamoylpyridine
4-(3-methylanilino)pyridine-3-sulfonamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents